Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Z-D-Abu-OH 98%

Supplier: AMBEED, INC
Warning

A224832-10G A224832-250MG A224832-25G A224832-5G
76722-080EA 17 USD InStock
76722-080 76722-082 76722-084 76722-086 76722-088
Z-D-Abu-OH 98%
Z-D-Abu-OH
Precaución: for Research Use Only


Formula: C₁₂H₁₅NO₄
MDL Number: MFCD00136772
CAS Number: 2900-20-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty