Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-hydroxy-2-naphthoate 97%

Supplier: AMBEED, INC
Warning

A227105-100G A227105-10G A227105-1G A227105-250MG A227105-25G A227105-5G
76722-142EA 187 USD InStock
76722-142 76722-144 76722-146 76722-148 76722-150 76722-152
Methyl-6-hydroxy-2-naphthoate 97%
Methyl 6-hydroxy-2-naphthoate
Precaución: for Research Use Only


Formula: C₁₂H₁₀O₃
MW: 202.2060 g/mol
MDL Number: MFCD00100526
CAS Number: 17295-11-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty