Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl methanesulfonylacetate 97%

Supplier: AMBEED, INC
Warning

A227611-100G A227611-10G A227611-1G A227611-25G A227611-5G
76722-164EA 66 USD InStock
76722-164 76722-166 76722-168 76722-170 76722-172
Methyl methanesulfonylacetate 97%
Methyl methanesulfonylacetate
Precaución: for Research Use Only


Formula: C₄H₈O₄S
MDL Number: MFCD00051842
CAS Number: 62020-09-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty