Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2-Pyridylmethyl)glycine ethyl ester 95%

Supplier: AMBEED, INC
Warning

A232369-1G A232369-5G
76722-262EA 30 USD InStock
76722-262 76722-264
N-(2-Pyridylmethyl)glycine ethyl ester 95%
N-(2-Pyridylmethyl)glycine ethyl ester
Precaución: for Research Use Only


Formula: C₁₀H₁₄N₂O₂
MDL Number: MFCD00191573
CAS Number: 62402-24-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty