Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-(chloromethyl)benzoate 95%

Supplier: AMBEED, INC
Danger

A238765-5G A238765-25G
76722-392EA 80 USD InStock
76722-392 77667-958
Methyl-2-(chloromethyl)benzoate 95%
Methyl 2-(chloromethyl)benzoate
Precaución: for Research Use Only


Formula: C₉H₉ClO₂
MW: 184.6196 g/mol
MDL Number: MFCD06200781
CAS Number: 34040-62-5
UN: 3265
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty