Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium butyrate 95%

Supplier: AMBEED, INC
Warning

A239433-100G A239433-25G A239433-500G A239433-5G A239433-1000G
76722-404EA 30 USD InStock
76722-404 76722-406 76722-408 76722-410 77667-968
Sodium butyrate 95%
Sodium butyrate
Precaución: for Research Use Only


Formula: C₄H₇NaO₂
MDL Number: MFCD00002816
CAS Number: 156-54-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty