Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 2-((2-aminoethyl)amino)ethanesulfonate 50% in water

Supplier: AMBEED, INC
Warning

A240611-100G A240611-10G A240611-25G A240611-500G A240611-5G
76722-430EA 18 USD InStock
76722-430 76722-432 76722-434 76722-436 76722-438
Sodium 2-((2-aminoethyl)amino)ethanesulfonate 50% in water
Sodium 2-((2-aminoethyl)amino)ethanesulfonate
Precaución: for Research Use Only


Formula: C₄H₁₁N₂NaO₃S
MDL Number: MFCD09838703
CAS Number: 34730-59-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty