Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Butylidenephthalide 97% (mixture of isomers)

Supplier: AMBEED, INC
Warning

A244960-100G A244960-25G A244960-5G
76722-512EA 100 USD InStock
76722-512 76722-514 76722-516
3-Butylidenephthalide 97% (mixture of isomers)
3-Butylidenephthalide
Precaución: for Research Use Only


Formula: C₁₂H₁₂O₂
MDL Number: MFCD00047319
CAS Number: 551-08-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty