Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Oxyclozanide 98%

Supplier: AMBEED, INC
Danger

A250572-10G A250572-1G A250572-5G
76722-606EA 5.31 USD InStock
76722-606 76722-608 76722-610 76722-612
Oxyclozanide 98%
Oxyclozanide
Precaución: for Research Use Only


Formula: C₁₃H₆Cl₅NO₃
MDL Number: MFCD00864507
CAS Number: 2277-92-1
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty