Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Diispropylbenzamide 95%

Supplier: AMBEED, INC
Warning

A256189-100MG A256189-1G A256189-250MG A256189-5G
76722-672EA 12 USD InStock
76722-672 76722-674 76722-676 76722-678
N,N-Diispropylbenzamide 95%
N,N-Diispropylbenzamide
Precaución: for Research Use Only


Formula: C₁₃H₁₉NO
MDL Number: MFCD00026370
CAS Number: 20383-28-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty