Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-p-toluate 97%

Supplier: AMBEED, INC
Warning

A258493-1000G A258493-100G A258493-10G A258493-25G A258493-500G
76722-730EA 51 USD InStock
76722-730 76722-732 76722-734 76722-736 76722-738
Methyl-p-toluate 97%
Methyl-p-toluate
Precaución: for Research Use Only


Formula: C₉H₁₀O₂
MW: 150.18
Boiling Pt: 217
Density: 1.06
Flash Pt: 89
MDL Number: MFCD00008441
CAS Number: 99-75-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty