Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Diphenylmethylene)-1,1-diethyl-2-methylpyrrolidin-1-ium bromide 97%

Supplier: AMBEED, INC
Warning

A259615-1G A259615-250MG A259615-5G
76722-760EA 33 USD InStock
76722-760 76722-762 76722-764
3-(Diphenylmethylene)-1,1-diethyl-2-methylpyrrolidin-1-ium bromide 97%
3-(Diphenylmethylene)-1,1-diethyl-2-methylpyrrolidin-1-ium bromide
Precaución: for Research Use Only


Formula: C₂₂H₂₈BrN
MW: 386.36 g/mol
MDL Number: MFCD00865211
CAS Number: 4630-95-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty