Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methylisonicotinate-N-oxide 95%

Supplier: AMBEED, INC
Warning

A263053-1G A263053-25G A263053-5G
76722-866EA 11 USD InStock
76722-866 76722-868 76722-870
Methylisonicotinate-N-oxide 95%
Methylisonicotinate-N-oxide
Precaución: for Research Use Only


Formula: C₇H₇NO₃
MW: 153.14 g/mol
MDL Number: MFCD00128856
CAS Number: 3783-38-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty