Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium dicyanoamide 98%

Supplier: AMBEED, INC
Warning

A263858-100G A263858-25G A263858-500G
76722-898EA 30 USD InStock
76722-898 76722-900 76722-902
Sodium dicyanoamide 98%
Sodium dicyanoamide
Precaución: for Research Use Only


Formula: C₂N₃Na
MDL Number: MFCD00003535
CAS Number: 1934-75-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty