Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Quinacrine dihydrochloride hydrate 98%

Supplier: AMBEED, INC
Warning

A266175-1G A266175-1MG
76722-950EA 57 USD InStock
76722-950 76722-952
Quinacrine dihydrochloride hydrate 98%
Quinacrine dihydrochloride hydrate
Precaución: for Research Use Only


Formula: C₂₃H₃₀ClN₃O
MDL Number: MFCD00012659
CAS Number: 69-05-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty