Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-bromoimidazo[1,2-a]pyridine-6-carboxylate 98%

Supplier: AMBEED, INC
Warning

A268770-100MG A268770-1G A268770-250MG A268770-5G
76722-984EA 8 USD InStock
76722-984 76722-986 76722-988 76722-990
Methyl-3-bromoimidazo[1,2-a]pyridine-6-carboxylate 98%
Methyl 3-bromoimidazo[1,2-a]pyridine-6-carboxylate
Precaución: for Research Use Only


Formula: C₉H₇BrN₂O₂
MDL Number: MFCD07021501
CAS Number: 886361-98-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty