Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-bromo-4-hydroxy-5-nitrobenzenecarboxylate 97%

Supplier: AMBEED, INC
Danger

A269175-10G A269175-1G A269175-250MG A269175-5G
76723-012EA 90 USD InStock
76723-012 76723-014 76723-016 76723-018
Methyl-3-bromo-4-hydroxy-5-nitrobenzenecarboxylate 97%
Methyl-3-bromo-4-hydroxy-5-nitrobenzenecarboxylate
Precaución: for Research Use Only


Formula: C₈H₆BrNO₅
MW: 276.05 g/mol
MDL Number: MFCD08689690
CAS Number: 40258-72-8
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty