Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-fluoroacrylate 97% stabilized

Supplier: AMBEED, INC
Danger

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

A272348-10G A272348-25G A272348-5G
76723-054EA 53 USD InStock
76723-054 76723-056 76723-058 76723-060
Methyl-2-fluoroacrylate 97% stabilized
Methyl-2-fluoroacrylate
Precaución: for Research Use Only


Formula: C₄H₅FO₂
MDL Number: MFCD04039286
CAS Number: 2343-89-7
UN: 1992
ADR: 3,6.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty