Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cinnamoyl-3-methylquinoxaline 1,4-dioxide 97%

Supplier: AMBEED, INC
Warning

A283041-25G A283041-25MG A283041-5G
76723-320EA 15 USD InStock
76723-320 76723-322 76723-324
2-Cinnamoyl-3-methylquinoxaline 1,4-dioxide 97%
2-Cinnamoyl-3-methylquinoxaline 1,4-dioxide
Precaución: for Research Use Only


Formula: C₁₈H₁₄N₂O₃
MDL Number: MFCD09753267
CAS Number: 81810-66-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty