Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexylamine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A283112-100G A283112-25G A283112-5G
76723-326EA 298 USD InStock
76723-326 76723-328 76723-330
Hexylamine hydrochloride 98%
Hexylamine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₅N
MW: 137.65 g/mol
MDL Number: MFCD00043792
CAS Number: 142-81-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty