Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Pyrrolidinone hydrotribromide 98%

Supplier: AMBEED, INC
Danger

A285169-100G A285169-25G A285169-5G
76723-384EA 64 USD InStock
76723-384 76723-386 76723-388
2-Pyrrolidinone hydrotribromide 98%
2-Pyrrolidinone hydrotribromide
Precaución: for Research Use Only


Formula: C₁₂H₂₂Br₃N₃O₃
MDL Number: MFCD00082241
CAS Number: 22580-55-8
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty