Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexyl nicotinate 98%

Supplier: AMBEED, INC
Warning

A288971-100G A288971-25G
76723-454EA 183 USD InStock
76723-454 76723-456
Hexyl nicotinate 98%
Hexyl nicotinate
Precaución: for Research Use Only


Formula: C₁₂H₁₇NO₂
MW: 207.27 g/mol
MDL Number: MFCD00023587
CAS Number: 23597-82-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty