Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyridin-4-ylacetonitrile 98%

Supplier: AMBEED, INC
Warning

A292957-1G A292957-250MG A292957-25G A292957-5G
76723-512EA 21 USD InStock
76723-512 76723-514 76723-516 76723-518
Pyridin-4-ylacetonitrile 98%
Pyridin-4-ylacetonitrile
Precaución: for Research Use Only


Formula: C₇H₆N₂
MDL Number: MFCD03412601
CAS Number: 13121-99-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty