Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 2-((4-(diethylamino)phenyl)(4-(diethyliminio)cyclohexa-2,5-dien-1-ylidene)methyl)benzene-1,4-disulfonate ≥99%

Supplier: AMBEED, INC
Warning

A295767-10MG A295767-1MG A295767-5MG
76723-562EA 27 USD InStock
76723-562 76723-564 76723-566
Sodium 2-((4-(diethylamino)phenyl)(4-(diethyliminio)cyclohexa-2,5-dien-1-ylidene)methyl)benzene-1,4-disulfonate ≥99%
Sodium 2-((4-(diethylamino)phenyl)(4-(diethyliminio)cyclohexa-2,5-dien-1-ylidene)methyl)benzene-1,4-disulfonate
Precaución: for Research Use Only


Formula: C₂₇H₃₁N₂NaO₆S₂
MDL Number: MFCD00867653
CAS Number: 68238-36-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty