Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium methyl sulphate 98%

Supplier: AMBEED, INC
Warning

A301019-100G A301019-5G
76723-622EA 293 USD InStock
76723-622 76723-624
Sodium methyl sulphate 98%
Sodium methyl sulphate
Precaución: for Research Use Only


Formula: CH₃NaO₄S
MW: 134.09 g/mol
Melting Pt: 193
MDL Number: MFCD00013457
CAS Number: 512-42-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty