Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-β-D-galactopyranoside 98%

Supplier: AMBEED, INC
Warning

A302833-100G A302833-5G A302833-25G
76723-638EA 248 USD InStock
76723-638 76723-640 76723-642 77601-362
Methyl-β-D-galactopyranoside 98%
Methyl-beta-D-galactopyranoside
Precaución: for Research Use Only


Formula: C₇H₁₄O₆
MDL Number: MFCD00064357
CAS Number: 1824-94-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty