Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pentachlorocyclopropane 94%

Supplier: AMBEED, INC
Warning

A305634-1G A305634-25G A305634-5G
76723-704EA 13 USD InStock
76723-704 76723-706 76723-708
Pentachlorocyclopropane 94%
Pentachlorocyclopropane
Precaución: for Research Use Only


Formula: C₃HCl₅
MDL Number: MFCD00010796
CAS Number: 6262-51-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty