Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium-3-(benzyloxy)phenyltrifluoroborate 97%

Supplier: AMBEED, INC
Warning

A306190-10G A306190-1G A306190-5G
76723-718EA 57 USD InStock
76723-718 76723-720 76723-722
Potassium-3-(benzyloxy)phenyltrifluoroborate 97%
Potassium 3-(benzyloxy)phenyltrifluoroborate
Precaución: for Research Use Only


Formula: C₁₃H₁₁BF₃KO
MW: 290.1300 g/mol
MDL Number: MFCD04115751
CAS Number: 850623-58-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty