Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexadecyltrimethylammonium tetrafluoroborate 98%

Supplier: AMBEED, INC
Danger

A306197-1G A306197-250MG A306197-25G A306197-5G
76723-724EA 45 USD InStock
76723-724 76723-726 76723-728 76723-730
Hexadecyltrimethylammonium tetrafluoroborate 98%
Hexadecyltrimethylammonium tetrafluoroborate
Precaución: for Research Use Only


Formula: C₁₉H₄₂BF₄N
MDL Number: MFCD00144896
CAS Number: 73257-08-6
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty