Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl (3-methylbenzyl)phosphonate ≥95%

Supplier: AMBEED, INC
Warning

A312020-25G A312020-5G
76723-838EA 36 USD InStock
76723-838 76723-840
Diethyl (3-methylbenzyl)phosphonate ≥95%
Diethyl (3-methylbenzyl)phosphonate
Precaución: for Research Use Only


Formula: C₁₂H₁₉O₃P
MW: 242.2512 g/mol
MDL Number: MFCD08276364
CAS Number: 63909-50-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty