Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Carbobenzoxy-D-threonine 98%

Supplier: AMBEED, INC
Warning

A315720-100G A315720-1G A315720-5G A315720-10G
76723-878EA 543 USD InStock
76723-878 76723-880 76723-882 77668-390
N-Carbobenzoxy-D-threonine 98%
N-Carbobenzoxy-D-threonine
Precaución: for Research Use Only


Formula: C₁₂H₁₅NO₅
MW: 253.25 g/mol
MDL Number: MFCD00037808
CAS Number: 80384-27-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty