Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-chloro-6-fluoro-2-(trifluoromethyl)quinazoline 95%

Supplier: AMBEED, INC
Warning

A317907-100MG A317907-1G
76723-912EA 36 USD InStock
76723-912 77668-414
4-chloro-6-fluoro-2-(trifluoromethyl)quinazoline 95%
4-chloro-6-fluoro-2-(trifluoromethyl)quinazoline
Precaución: for Research Use Only


Formula: C₉H₃ClF₄N₂
MW: 250.58 g/mol
MDL Number: MFCD09954902
CAS Number: 959238-11-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty