Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenoxybenzaldehyde 98%

Supplier: AMBEED, INC
Warning

A318600-100G A318600-1G A318600-25G A318600-5G
76723-924EA 571 USD InStock
76723-924 76723-926 76723-928 76723-930
2-Phenoxybenzaldehyde 98%
2-Phenoxybenzaldehyde
Precaución: for Research Use Only


Formula: C₁₃H₁₀O₂
MDL Number: MFCD00800666
CAS Number: 19434-34-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty