Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Chloroethyl)-1H-isoindole-1,3(2H)-dione 95%

Supplier: AMBEED, INC
Warning

A320681-250MG A320681-25G A320681-5G
76723-966EA 5 USD InStock
76723-966 76723-968 76723-970
2-(2-Chloroethyl)-1H-isoindole-1,3(2H)-dione 95%
2-(2-Chloroethyl)-1H-isoindole-1,3(2H)-dione
Precaución: for Research Use Only


Formula: C₁₀H₈ClNO₂
MDL Number: MFCD00129356
CAS Number: 6270-06-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty