Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Mesitylenesulfonyl)imidazole 98%

Supplier: AMBEED, INC
Warning

A329295-100G A329295-10G A329295-1G A329295-25G A329295-5G
76724-112EA 80 USD InStock
76724-112 76724-114 76724-116 76724-118 76724-120
1-(2-Mesitylenesulfonyl)imidazole 98%
1-(2-Mesitylenesulfonyl)imidazole
Precaución: for Research Use Only


Formula: C₁₂H₁₄N₂O₂S
MDL Number: MFCD00005284
CAS Number: 50257-39-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty