Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ɑ-Cyclopentyl-DL-mandelic acid 97%

Supplier: AMBEED, INC
Danger

A329412-10G A329412-1G A329412-25G A329412-5G
76724-122EA 13 USD InStock
76724-122 76724-124 76724-126 76724-128
ɑ-Cyclopentyl-DL-mandelic acid 97%
alpha-Cyclopentyl-DL-mandelic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₆O₃
MW: 220.27 g/mol
MDL Number: MFCD00019296
CAS Number: 427-49-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty