Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-nitrobenzoate 98%

Supplier: AMBEED, INC
Warning

A334902-100G A334902-25G A334902-5G
76724-212EA 77 USD InStock
76724-212 76724-214 76724-216
Methyl-2-nitrobenzoate 98%
Methyl-2-nitrobenzoate
Precaución: for Research Use Only


Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 268
Melting Pt: -13
MDL Number: MFCD00007136
CAS Number: 606-27-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty