Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Salicylohydroxamic acid 97%

Supplier: AMBEED, INC
Warning

A335264-100G A335264-10G A335264-1G A335264-500G A335264-5G A335264-25G
76724-226EA 83 USD InStock
76724-226 76724-228 76724-230 76724-232 76724-234 77306-484
Salicylohydroxamic acid 97%
Salicylohydroxamic acid
Precaución: for Research Use Only


Formula: C₇H₇NO₃
MDL Number: MFCD00002110
CAS Number: 89-73-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty