Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl quinoxaline-6-carboxylate 98%

Supplier: AMBEED, INC
Warning

A340582-1G A340582-5G
76724-330EA 20 USD InStock
76724-330 76724-332
Methyl quinoxaline-6-carboxylate 98%
Methyl quinoxaline-6-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₈N₂O₂
MDL Number: MFCD00102618
CAS Number: 23088-23-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty