Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium (piperazin-1-yl)methyltrifluoroborate, N-4 BOC protected 98%

Supplier: AMBEED, INC
Warning

A341048-1G A341048-25G A341048-5G
76724-334EA 16 USD InStock
76724-334 76724-336 76724-338
Potassium (piperazin-1-yl)methyltrifluoroborate, N-4 BOC protected 98%
Potassium (piperazin-1-yl)methyltrifluoroborate, N-4 BOC protected
Precaución: for Research Use Only


Formula: C₁₀H₁₉BF₃N₂O₂·K
MW: 306.175 g/mol
MDL Number: MFCD10566514
CAS Number: 936329-97-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty