Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Aminophenyl)benzonitrile 98%

Supplier: AMBEED, INC
Danger

A341338-100MG A341338-1G A341338-250MG A341338-5G
76724-340EA 12 USD InStock
76724-340 76724-342 76724-344 76724-346
4-(4-Aminophenyl)benzonitrile 98%
4-(4-Aminophenyl)benzonitrile
Precaución: for Research Use Only


Formula: C₁₃H₁₀N₂
MDL Number: MFCD00191367
CAS Number: 4854-84-6
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty