Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl phenylsulfonylacetate 98%

Supplier: AMBEED, INC
Warning

A351507-100G A351507-1G A351507-25G A351507-5G
76724-466EA 128 USD InStock
76724-466 76724-468 76724-470 76724-472
Ethyl phenylsulfonylacetate 98%
Ethyl phenylsulfonylacetate
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₄S
MDL Number: MFCD00025041
CAS Number: 7605-30-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty