Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyridine trifluoroacetate 98%

Supplier: AMBEED, INC
Warning

A359728-100G A359728-1G A359728-25G A359728-5G
76724-564EA 37 USD InStock
76724-564 76724-566 76724-568 76724-570
Pyridine trifluoroacetate 98%
Pyridine trifluoroacetate
Precaución: for Research Use Only


Formula: CF₃CO₂H·C₅H₅N
MW: 193.123 g/mol
MDL Number: MFCD00013072
CAS Number: 464-05-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty