Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-tert-Butylphthalic anhydride 95%

Supplier: AMBEED, INC
Warning

A364535-1G A364535-250MG A364535-25G A364535-5G
76724-634EA 23 USD InStock
76724-634 76724-636 76724-638 76724-640
4-tert-Butylphthalic anhydride 95%
4-tert-Butylphthalic anhydride
Precaución: for Research Use Only


Formula: C₁₂H₁₂O₃
MW: 204.2219 g/mol
MDL Number: MFCD00060127
CAS Number: 32703-79-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty