Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Diethylacetoacetamide 98%

Supplier: AMBEED, INC
Warning

A370008-100G A370008-25G A370008-5G
76724-738EA 55 USD InStock
76724-738 76724-740 76724-742 76724-744
N,N-Diethylacetoacetamide 98%
N,N-Diethylacetoacetamide
Precaución: for Research Use Only


Formula: C₈H₁₅NO₂
MW: 157.20 g/mol
MDL Number: MFCD00026728
CAS Number: 2235-46-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty