Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pentafluorophenyl acrylate 98% stabilized

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with 4-methoxyphenol (MEHQ) 500 ppm

A384484-250MG A384484-25G A384484-5G
76724-904EA 9 USD InStock
76724-904 76724-906 77551-128
Pentafluorophenyl acrylate 98% stabilized
Pentafluorophenyl acrylate
Precaución: for Research Use Only


Formula: C₉H₃F₅O₂
MDL Number: MFCD00042330
CAS Number: 71195-85-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty