Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl (4-piperidyl)acetate hydrochloride 98%

Supplier: AMBEED, INC
Warning

A385225-10G A385225-1G A385225-25G A385225-5G
76724-916EA 34 USD InStock
76724-916 76724-918 76724-920 76724-922 76724-924
Methyl (4-piperidyl)acetate hydrochloride 98%
Methyl (4-piperidyl)acetate hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₅NO₂
MDL Number: MFCD09261419
CAS Number: 81270-37-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty