Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenyl-p-toluenesulfonate 99%

Supplier: AMBEED, INC
Warning

A386049-100G A386049-10G A386049-1G A386049-5G
76724-932EA 161 USD InStock
76724-932 76724-934 76724-936 76724-938
Phenyl-p-toluenesulfonate 99%
Phenyl-p-toluenesulfonate
Precaución: for Research Use Only


Formula: C₁₃H₁₂O₃S
MDL Number: MFCD00025996
CAS Number: 640-60-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty