Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3R,3AS,6R,7R,8aS)-3-methoxy-3,6,8,8-tetramethyloctahydro-1H-3a,7-methanoazulene 96%

Supplier: AMBEED, INC
Warning

A403473-1000G A403473-500G
76725-108EA 179 USD InStock
76725-108 76725-110
(3R,3AS,6R,7R,8aS)-3-methoxy-3,6,8,8-tetramethyloctahydro-1H-3a,7-methanoazulene 96%
(3R,3AS,6R,7R,8aS)-3-methoxy-3,6,8,8-tetramethyloctahydro-1H-3a,7-methanoazulene
Precaución: for Research Use Only


Formula: C₁₆H₂₈O
MDL Number: MFCD00216983
CAS Number: 19870-74-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty