Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-chloro-2'-fluorobenzophenone 98%

Supplier: AMBEED, INC
Warning

A414668-100G A414668-10G A414668-500G A414668-5G A414668-25G
76725-198EA 53 USD InStock
76725-198 76725-200 76725-202 76725-204 76841-896
2-Amino-5-chloro-2'-fluorobenzophenone 98%
2-Amino-5-chloro-2'-fluorobenzophenone
Precaución: for Research Use Only


Formula: C₁₃H₉ClFNO
MW: 249.67 g/mol
Boiling Pt: 364
MDL Number: MFCD00038381
CAS Number: 784-38-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty